siRNA Details
Gene Symbol: Dennd4c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_184088.1 | NP_908976.1 | 17 | 2660 | ||
GenBank mRNA | AK155220.1 | - | 17 | 2661 | ||
BC146602.1 | - | 16 | 2285 | 40099465 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
