siRNA Details
Gene Symbol: Acot11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025590.4 | NP_079866.2 | 16 | 1787 | ||
GenBank mRNA | AF416923.1 | - | 16 | 1725 | ||
AK031588.1 | - | 16 | 1704 | |||
AK038546.1 | - | 16 | 1725 | |||
AK048670.1 | - | 16 | 1787 | |||
AK052332.1 | - | 16 | 1803 | |||
AK122350.1 | - | 15 | 1574 | |||
AK170189.1 | - | 9 | 810 | |||
AK171255.1 | - | 9 | 811 | |||
BC042492.1 | - | 16 | 1722 | 4240966 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Rat: NM_001271383, XM_008763852, XM_008763851.
Gene Ontology Categories:
