siRNA Details
Gene Symbol: Acot11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025590.4 | NP_079866.2 | 15, 16 | 1742 | ||
GenBank mRNA | AF416923.1 | - | 15, 16 | 1680 | ||
AK031588.1 | - | 15, 16 | 1659 | |||
AK038546.1 | - | 15, 16 | 1680 | |||
AK048670.1 | - | 15, 16 | 1742 | |||
AK052332.1 | - | 15, 16 | 1758 | |||
AK122350.1 | - | 14, 15 | 1529 | |||
AK170189.1 | - | 8, 9 | 765 | |||
AK171255.1 | - | 8, 9 | 766 | |||
BC042492.1 | - | 15, 16 | 1677 | 4240966 |
