siRNA Details
Gene Symbol: Fhad1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177868.4 | NP_808536.2 | 20 | 2774 | ||
GenBank mRNA | AK148490.1 | - | 5 | 859 | ||
BC038692.1 | - | 2 | 357 | 1496088 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Fhad1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177868.4 | NP_808536.2 | 20 | 2774 | ||
GenBank mRNA | AK148490.1 | - | 5 | 859 | ||
BC038692.1 | - | 2 | 357 | 1496088 |