siRNA Details
Gene Symbol: Slc5a6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001177621.1 | NP_001171092.1 | 11, 12 | 1862 | ||
NM_001177622.1 | NP_001171093.1 | 11, 12 | 1644 | |||
NM_177870.5 | NP_808538.1 | 10, 11 | 1751 | |||
GenBank mRNA | AK088682.1 | - | 11, 12 | 1550 | ||
AK158896.1 | - | 11, 12 | 1863 | |||
AK161633.1 | - | 10, 11 | 1675 | |||
AY572835.1 | - | 8, 9 | 1102 | |||
BC065061.1 | - | 11, 12 | 1644 | |||
BC085132.1 | - | 10, 11 | 1447 | 6416422 | ||
BC117852.1 | - | 10, 11 | 1404 | 40094012 | ||
BC117853.1 | - | 10, 11 | 1404 | 40094013 |
