siRNA Details
Gene Symbol: Shisa3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033415.3 | NP_001028587.1 | 2 | 1128 | ||
GenBank mRNA | AK077061.1 | - | 2 | 369 | ||
AK143290.1 | - | 2 | 1129 | |||
BC120534.1 | - | 2 | 636 | 40129457 | ||
BC120536.1 | - | 2 | 636 | 40129459 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
