siRNA Details
Gene Symbol: RAB1A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004161.4 | NP_004152.1 | 4, 5 | 677 | ||
GenBank mRNA | AF498929.1 | - | 4, 5 | 291 | ||
AK315495.1 | - | 4, 5 | 486 | |||
BC000905.2 | - | 4, 5 | 469 | 2900705 | ||
BT019528.1 | - | 4, 5 | 291 | |||
BT019529.1 | - | 4, 5 | 291 | |||
BX571747.1 | - | 4, 5 | 474 | |||
CR536488.1 | - | 4, 5 | 291 | |||
M28209.1 | - | 4, 5 | 341 | |||
X13898.1 | - | 4, 5 | 291 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
