siRNA Details
Gene Symbol: Kctd10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001159941.1 | NP_001153413.1 | 6 | 623 | ||
NM_026145.4 | NP_080421.2 | 6 | 620 | |||
GenBank mRNA | AK028657.1 | - | 6 | 587 | ||
AK041516.1 | - | 6 | 587 | |||
AK145295.1 | - | 6 | 599 | |||
AK147714.1 | - | 6 | 565 | |||
AK161199.1 | - | 6 | 577 | |||
AK163351.1 | - | 6 | 574 | |||
AK166505.1 | - | 6 | 599 | |||
AK216751.1 | - | 3 | 113 | |||
BC006935.1 | - | 6 | 601 | 3601917 | ||
BQ571808.1 | - | 6 | 574 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
