siRNA Details
Gene Symbol: Ccdc63 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001289809.1 | NP_001276738.1 | 10, 11 | 1623 | ||
NM_001289811.1 | NP_001276740.1 | 11, 12 | 1743 | |||
NM_183307.4 | NP_899130.1 | 11, 12 | 1885 | |||
GenBank mRNA | AK029513.1 | - | 11, 12 | 1887 | ||
BC118944.1 | - | 10, 11 | 1545 | |||
BC118945.1 | - | 11, 12 | 1665 | 40092360 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
