siRNA Details
Gene Symbol: 4933411G11Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177880.4 | NP_808548.1 | 7 | 1179 | ||
GenBank mRNA | AK030175.1 | - | 7 | 1180 | ||
BC115568.1 | - | 7 | 712 | 40102090 | ||
BC115569.1 | - | 7 | 801 | 40102091 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
