siRNA Details
Gene Symbol: Gm5111 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_183309.3 | NP_899132.1 | 2 | 379 | ||
GenBank mRNA | AK041789.1 | - | 2 | 379 | ||
BC104366.1 | - | 2 | 374 | |||
BC104367.1 | - | 1 | 408 | |||
BC127138.1 | - | 1 | 410 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
