siRNA Details
Gene Symbol: Gm765 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001128092.1 | NP_001121564.1 | 3 | 404 | ||
GenBank mRNA | BC147266.1 | - | 3 | 404 | 9056347 | |
BC147267.1 | - | 3 | 404 | 9056335 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
