siRNA Details
Gene Symbol: Tmem52b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081186.1 | NP_001074655.1 | 4 | 746 | ||
GenBank mRNA | AK085389.1 | - | 4 | 750 | ||
AK085571.1 | - | 4 | 687 | |||
BC120661.1 | - | 4 | 399 | 40129584 | ||
BC120663.1 | - | 4 | 399 | 40129586 | ||
BC144939.1 | - | 3 | 355 |

More Information
Other Species Targeted

Gene Ontology Categories:
