siRNA Details
Gene Symbol: Gm766 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145390.1 | NP_001138862.1 | 1 | 174 | ||
GenBank mRNA | AK144423.1 | - | 211 | |||
BC151094.1 | - | 1 | 174 | 9088007 | ||
BC151096.1 | - | 1 | 174 | 9088009 | ||
BC172155.1 | - | 1 | 174 | 9054825 |
