siRNA Details
Gene Symbol: Tmem150b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001142792.1 | NP_001136264.1 | 2, 3 | 228 | ||
NM_177887.4 | NP_808555.1 | 2, 3 | 397 | |||
GenBank mRNA | AK041846.1 | - | 2, 3 | 398 | ||
BC022651.1 | - | 2, 3 | 228 | 4216812 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
