siRNA Details
Gene Symbol: Bsph1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033418.4 | NP_001028590.1 | 2, 3 | 166 | ||
GenBank mRNA | AK136662.1 | - | 2, 3 | 167 | ||
BC147454.1 | - | 2, 3 | 151 | 9056096 | ||
BC147459.1 | - | 2, 3 | 151 | 9056101 | ||
DQ227498.1 | - | 2, 3 | 146 |
