siRNA Details
Gene Symbol: Nlrp9c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001042612.1 | NP_001036077.1 | 2 | 381 | ||
GenBank mRNA | AK163321.1 | - | 2 | 149 | ||
AY360472.1 | - | 376 | ||||
AY596203.1 | - | 2 | 381 |
