siRNA Details
Gene Symbol: Gm5113 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033540.2 | NP_001028712.1 | 3 | 1496 | ||
GenBank mRNA | AK046598.1 | - | 3 | 1545 | ||
AK142560.1 | - | 3 | 1282 | |||
AK147880.1 | - | 3 | 1497 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
