siRNA Details
Gene Symbol: Prss53 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081268.1 | NP_001074737.1 | 4 | 413 | ||
GenBank mRNA | AK220244.1 | - | 4 | 370 | ||
BC147509.1 | - | 4 | 410 | 9056457 | ||
BC151010.1 | - | 4 | 410 | 9087923 | ||
BC151017.1 | - | 4 | 410 | 9087930 | ||
BC172059.1 | - | 4 | 410 | 9054345 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
