siRNA Details
Gene Symbol: Btbd16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081038.2 | NP_001074507.2 | 5, 6 | 787 | ||
GenBank mRNA | AK087912.1 | - | 5, 6 | 789 | ||
BB559682.2 | - | 3, 4 | 346 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
