siRNA Details
Gene Symbol: Snapc5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_183316.2 | NP_899139.2 | 1 | 103 | ||
GenBank mRNA | BC028529.1 | - | 1 | 92 | 1381169 | |
CJ165571.1 | - | 1 | 104 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
