siRNA Details
Gene Symbol: Slc9a9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177909.5 | NP_808577.3 | 2 | 472 | ||
GenBank mRNA | AK037058.1 | - | 2 | 456 | ||
AK156494.1 | - | 2 | 433 | |||
AK172687.1 | - | 2 | 444 | |||
BC099942.1 | - | 2 | 446 | 30103308 | ||
BC153825.1 | - | 2 | 450 | 40093280 |
