siRNA Details
Gene Symbol: Gm773 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033423.2 | NP_001028595.1 | 3, 4 | 248 | ||
GenBank mRNA | AK144779.1 | - | 3, 4 | 249 | ||
AK165870.1 | - | 3, 4 | 246 | |||
BC147685.1 | - | 2, 3 | 179 | 9056658 | ||
BC147688.1 | - | 2, 3 | 179 | 9056661 |
