siRNA Details
Gene Symbol: Uprt | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081189.1 | NP_001074658.1 | 5 | 778 | ||
GenBank mRNA | BC147845.1 | - | 5 | 636 | 9087562 | |
BC147853.1 | - | 5 | 636 | 9087570 |

More Information
Other Species Targeted

Gene Ontology Categories:
