siRNA Details
Gene Symbol: Nanos1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178421.3 | NP_848508.2 | 1 | 986 | ||
GenBank mRNA | AB095029.1 | - | 1 | 564 | ||
AK046189.1 | - | 1 | 589 | |||
AK139148.1 | - | 1 | 796 | |||
BC056473.1 | - | 1 | 673 | |||
BC085512.1 | - | 1 | 986 | 6849859 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
