siRNA Details
Gene Symbol: Tmem211 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033428.2 | NP_001028600.1 | 5 | 675 | ||
GenBank mRNA | AK138479.1 | - | 5 | 676 | ||
AK138629.1 | - | 5 | 676 | |||
BC147409.1 | - | 4 | 538 | 9056497 | ||
BC147410.1 | - | 4 | 538 | 9056496 | ||
BC172062.1 | - | 4 | 535 | 9054348 |
