siRNA Details
Gene Symbol: Ksr2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001114545.2 | NP_001108017.2 | 3 | 1285 | ||
GenBank mRNA | AK137433.1 | - | 3 | 1287 | ||
BC068233.1 | - | 1 | 70 | |||
BC150883.1 | - | 3 | 1285 | |||
BC150890.1 | - | 3 | 1285 | |||
DQ531035.1 | - | 3 | 458 |
