siRNA Details
Gene Symbol: 4930453H23Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001252013.1 | NP_001238942.1 | 5 | 1266 | ||
GenBank mRNA | AK015451.1 | - | 5 | 1268 |
Gene Symbol: Gm15104 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001101501.1 | NP_001094971.1 | 1 | 282 | ||
GenBank mRNA | BC099570.1 | - | 2 | 359 |

More Information
Other Species Targeted

Gene Ontology Categories:
