siRNA Details
Gene Symbol: Gm867 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001277132.1 | NP_001264061.1 | 3 | 193 | ||
GenBank mRNA | AK144847.1 | - | 3 | 194 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Gm867 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001277132.1 | NP_001264061.1 | 3 | 193 | ||
GenBank mRNA | AK144847.1 | - | 3 | 194 |