siRNA Details
Gene Symbol: Cntln | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175275.4 | NP_780484.2 | 5 | 900 | ||
NM_177385.4 | NP_796359.1 | 5 | 900 | |||
GenBank mRNA | AK031604.1 | - | 5 | 806 | ||
AK052558.1 | - | 5 | 820 | |||
AK165111.1 | - | 4 | 373 | |||
BC125622.1 | - | 5 | 775 | |||
BC125624.1 | - | 5 | 775 |
