siRNA Details
Gene Symbol: Map3k9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001174107.1 | NP_001167578.1 | 2, 3 | 1087 | ||
NM_177395.5 | NP_796369.2 | 2, 3 | 1087 | |||
GenBank mRNA | AK053843.1 | - | 2, 3 | 928 | ||
AK155677.1 | - | 2, 3 | 1093 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
