siRNA Details
Gene Symbol: Atp6v1g3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177397.3 | NP_796371.1 | 3 | 283 | ||
GenBank mRNA | AB088359.1 | - | 3 | 283 | ||
AK032887.1 | - | 3 | 283 | |||
BC107267.1 | - | 3 | 282 | 40052586 | ||
BC107268.1 | - | 3 | 282 | 40052587 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
