siRNA Details
Gene Symbol: Cndp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177450.4 | NP_803233.2 | 8 | 1137 | ||
GenBank mRNA | AK085308.1 | - | 8 | 1127 | ||
BC043305.1 | - | 8 | 1066 | 5040944 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
