siRNA Details
Gene Symbol: Kdm7a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033430.4 | NP_001028602.2 | 17 | 2379 | ||
GenBank mRNA | AK035529.1 | - | 8 | 1393 | ||
AK129429.1 | - | 6 | 721 | |||
AK136169.1 | - | 5 | 517 | |||
AK136209.1 | - | 17 | 2218 | |||
AK136238.1 | - | 17 | 2366 | |||
BC007161.1 | - | 17 | 2075 | |||
BC145216.1 | - | 17 | 2265 | |||
BC145848.1 | - | 17 | 2269 | 40131151 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
