siRNA Details
Gene Symbol: Pramel4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001001319.3 | NP_001001319.2 | 4 | 1527 | ||
GenBank mRNA | AF490340.1 | - | 4 | 1411 | ||
AK145585.1 | - | 4 | 1528 | |||
AK163305.1 | - | 4 | 1386 |
Gene Symbol: Gm13102 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001085419.1 | NP_001078888.1 | 4 | 1526 | ||
GenBank mRNA | BC132592.1 | - | 4 | 1527 | 40130869 | |
BC132620.1 | - | 4 | 1527 | 40130897 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
