siRNA Details
Gene Symbol: Egfl7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164564.1 | NP_001158036.1 | 7 | 720 | ||
NM_178444.4 | NP_848538.3 | 8 | 945 | |||
NM_198724.2 | NP_942017.2 | 8 | 944 | |||
NM_198725.2 | NP_942018.2 | 8 | 944 | |||
GenBank mRNA | AF184973.1 | - | 8 | 895 | ||
AK002601.1 | - | 8 | 940 | |||
AK139377.1 | - | 7 | 715 | |||
AK156158.1 | - | 8 | 947 | |||
AY239289.1 | - | 8 | 906 | |||
AY239290.1 | - | 8 | 928 | |||
AY309459.1 | - | 8 | 918 | |||
BC024610.1 | - | 8 | 928 | 4159675 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
