siRNA Details
Gene Symbol: Edc3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153799.3 | NP_722494.1 | 5, 6 | 1124 | ||
GenBank mRNA | AK090005.1 | - | 815 | |||
AK140127.1 | - | 4, 5 | 819 | |||
AK146804.1 | - | 5, 6 | 1154 | |||
AK166286.1 | - | 5, 6 | 1121 | |||
AK168118.1 | - | 5, 6 | 1132 | |||
BC031725.1 | - | 5, 6 | 1124 | 4017604 | ||
BC033484.1 | - | 5, 6 | 1125 | 4954674 |

More Information
Other Species Targeted

Gene Ontology Categories:
