siRNA Details
Gene Symbol: Defb37 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_181683.2 | NP_859011.1 | 1 | 127 | ||
GenBank mRNA | AJ489588.1 | - | 1 | 127 | ||
AJ578470.1 | - | 1 | 45 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
