siRNA Details
Gene Symbol: Cmtm1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AY241873.1 | - | 578 | |||
AY241874.1 | - | 1012 | ||||
AY369078.2 | - | 84 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
