siRNA Details
Gene Symbol: Defb38 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_183036.1 | NP_898857.1 | 2 | 116 | ||
GenBank mRNA | AJ575424.1 | - | 2 | 153 | ||
AJ578471.1 | - | 2 | 116 | |||
BC125388.1 | - | 2 | 160 | 40129903 | ||
BC125390.1 | - | 2 | 160 | 40129905 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
