siRNA Details
Gene Symbol: Serf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290837.1 | NP_001277766.1 | 2 | 293 | ||
NM_011354.3 | NP_035484.1 | 2 | 293 | |||
GenBank mRNA | AF073297.1 | - | 2 | 74 | ||
AK002351.1 | - | 2 | 69 | |||
AK008802.1 | - | 2 | 82 | |||
AK010749.1 | - | 2 | 125 | |||
AK012395.1 | - | 2 | 86 | |||
AK012603.1 | - | 2 | 72 | |||
AK018746.1 | - | 2 | 69 | |||
AK132746.1 | - | 2 | 86 | |||
AK151362.1 | - | 2 | 86 | |||
AK168858.1 | - | 2 | 73 | |||
AK169106.1 | - | 2 | 75 | |||
BC081455.1 | - | 2 | 77 | |||
BY070249.1 | - | 2 | 109 | |||
CJ073462.1 | - | 2 | 299 |

More Information
Other Species Targeted

Gene Ontology Categories:
