siRNA Details
Gene Symbol: Lrrc24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198119.2 | NP_932787.1 | 6 | 1602 | ||
GenBank mRNA | AK048678.1 | - | 6 | 1595 | ||
AK078176.1 | - | 6 | 1603 | |||
BC116884.1 | - | 4 | 1394 | 40126203 | ||
BC116886.1 | - | 4 | 1394 | 40126205 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
