siRNA Details
Gene Symbol: Fastkd5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001146084.1 | NP_001139556.1 | 3 | 1531 | ||
NM_198176.2 | NP_937819.2 | 3 | 1568 | |||
GenBank mRNA | AK143929.1 | - | 3 | 1507 | ||
AK220420.1 | - | 1 | 1132 | |||
BC048956.1 | - | 1 | 570 | |||
BC052898.1 | - | 3 | 1488 | 30059761 |
Gene Symbol: Ubox5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK170024.1 | - | 2 | 1426 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
