siRNA Details
Gene Symbol: Sec14l3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001029937.2 | NP_001025108.1 | 6 | 547 | ||
GenBank mRNA | BC138982.1 | - | 6 | 547 | 8862004 | |
BC138985.1 | - | 6 | 547 | 8862007 | ||
BC145485.1 | - | 6 | 590 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
