siRNA Details
Gene Symbol: Ccnjl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001045530.2 | NP_001038995.1 | 4 | 563 | ||
GenBank mRNA | AK143205.1 | - | 4 | 529 | ||
BC139344.1 | - | 3 | 324 | 8862366 | ||
BC139345.1 | - | 3 | 324 | 8862367 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
