siRNA Details
Gene Symbol: Tmem102 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033433.4 | NP_001028605.2 | 2 | 909 | ||
GenBank mRNA | AK143262.1 | - | 2 | 909 | ||
BC144871.1 | - | 1 | 959 |

More Information
Other Species Targeted

Gene Ontology Categories:
