siRNA Details
Gene Symbol: Spata22 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001045531.1 | NP_001038996.1 | 7 | 774 | ||
GenBank mRNA | BC147586.1 | - | 7 | 756 | 9056576 | |
BC147607.1 | - | 7 | 756 | 9056597 |
