siRNA Details
Gene Symbol: Rap1gap2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001015046.2 | NP_001015046.1 | 16 | 1449 | ||
GenBank mRNA | AK038834.1 | - | 6 | 512 | ||
AK122424.1 | - | 2 | 242 | |||
AK147290.1 | - | 17 | 1431 | |||
BC107388.2 | - | 7 | 655 | 40057754 |

More Information
Other Species Targeted

Gene Ontology Categories:
