siRNA Details
Gene Symbol: Rap1gap2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001015046.2 | NP_001015046.1 | 7, 8 | 639 | ||
GenBank mRNA | AK147290.1 | - | 8, 9 | 621 | ||
CF743736.1 | - | 7, 8 | 503 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
