siRNA Details
Gene Symbol: Scarf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001004157.2 | NP_001004157.2 | 10, 11 | 1676 | ||
GenBank mRNA | AK133477.1 | - | 10, 11 | 1659 | ||
AK220215.1 | - | 10, 11 | 1660 | |||
BC057600.1 | - | 9, 10 | 1583 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
